Learn More
Neogen Campylobacter Enrichment Broth CE250
Product Code. 17263784
Encompass
Change view
Click to view available options
Weight: 250 g
Product Code.
17263784 Weight: 250 g
1 options
Product Code. 17263784 Supplier Neogen Supplier No. 7100133299
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Aerobically incubated medium used for the selective enrichment of Campylobacter
  • For the selective enrichment of Campylobacter
  • Aerobic incubation - no need for a microaerophilic environment
  • Ready to Use enrichment - just add sterile water (no supplements or organic solvents)
  • Use with MDA2CAM96 Neogen Molecular Detection Assay 2 - Campylobacter for fast and reliable results.
  • 250 Gram bottle makes approximately 203 bird rinse enrichments
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Test Type Food and Beverage, Environmental Test
Quantity 1
Weight (Metric) 250 g