missing translation for 'onlineSavingsMsg'
Learn More

Camphor, MedChemExpress

Product Code. 16409146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409146 Supplier Medchem Express Supplier No. HYN0808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Camphor ((±)-Camphor) is a topical anti-infective and anti-pruritic and internally as a stimulant and carminative. However, Camphor is poisonous when ingested. Antiviral, antitussive, and anticancer activities. Camphor is a TRPV3 agonist.

Camphor ((±)-Camphor) is a topical anti-infective and anti-pruritic and internally as a stimulant and carminative. However, Camphor is poisonous when ingested. Antiviral, antitussive, and anticancer activities. Camphor is a TRPV3 agonist.

Specifications

Chemical Name or Material Camphor
Physical Form Solid
CAS 76-22-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H16O
For Use With (Application) Cancer-Kinase/protease
Synonym (±)-Camphor
Solubility Information DMSO : 100 mg/mL (656.90 mM; Need ultrasonic) ∣H2O : 5 mg/mL (32.85 mM; Need ultrasonic)
SMILES O=C1C(C2(C)C)(C)CCC2C1
Molecular Weight (g/mol) 152.23
Formula Weight 152.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.