Learn More
(±)-Camphor, 96%
Description
Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.
Solubility
Soluble in acetone, ethanol, diethylether, chloroform and acetic acid.
Notes
Incompatible with strong oxidizing agents, strong reducing agents and chlorinated solvents.
Specifications
Specifications
| Melting Point | 172°C to 176°C |
| Density | 0.992 g/mL |
| Boiling Point | 204°C |
| Flash Point | 64°C (147°F) |
| Assay Percent Range | 96% |
| Quantity | 500 g |
| UN Number | UN2717 |
| Beilstein | 1907611 |
| Merck Index | 14,1732 |
| Solubility Information | Soluble in acetone,ethanol,diethylether,chloroform and acetic acid. |
| Show More |
Safety and Handling
- (+/-)-Camphor
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 2
- H228-Flammable solid.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contain: (+/-)-Camphor
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.