missing translation for 'onlineSavingsMsg'
Learn More

(±)-Camphor, 96%

Product Code. 11478960
Change view
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478960 100 g 100g
11454343 500 g 500g
11464343 2500 g 2500g
11488960 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11478960 Supplier Thermo Scientific Alfa Aesar Supplier No. A10936.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-22-2 | C10H16O | 152.24 g/mol

Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.

Solubility
Soluble in acetone, ethanol, diethylether, chloroform and acetic acid.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and chlorinated solvents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-22-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074738,MFCD00064149
InChI Key DSSYKIVIOFKYAU-UHFFFAOYNA-N
Synonym camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
PubChem CID 2537
ChEBI CHEBI:36773
IUPAC Name 4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C)C2CCC1(C)C(=O)C2

Specifications

Melting Point 172°C to 176°C
Density 0.992
Boiling Point 204°C
Flash Point 64°C (147°F)
Assay Percent Range 96%
Odor Camphor-like
Quantity 100 g
UN Number UN2717
Beilstein 1907611
Merck Index 14,1732
Solubility Information Soluble in acetone,ethanol,diethylether,chloroform and acetic acid.
Formula Weight 152.24
Percent Purity 96%
Chemical Name or Material (±)-Camphor
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+/-)-Camphor
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: Camphor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.