missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphene, tech. (sum of camphene + fenchene)

Product Code. 11429186
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429186 50 g 50g
11439186 250 g 250g
11419186 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429186 Supplier Thermo Scientific Alfa Aesar Supplier No. A18040.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-92-5 | C10H16 | 136.24 g/mol

Camphene is used in the preparation of fragrances and as a food additive for flavoring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYNA-N

Specifications

Melting Point ∼35°C
Boiling Point 159°C to 160°C
Flash Point 34°C (94°F)
Quantity 50 g
UN Number UN1325
Merck Index 14,1730
Synonym 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane
SMILES CC1(C)C2CCC(C2)C1=C
IUPAC Name (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
PubChem CID 6560154
Formula Weight 136.24
Grade Technical
Chemical Name or Material (+/-)-Camphene, sum of camphene + fenchene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.