missing translation for 'onlineSavingsMsg'
Learn More

Camphene, 75%, remainder mainly alpha-fenchene

Product Code. p-82069 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10351054 1 L Glass bottle 1 L
10115800 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351054 Supplier Thermo Scientific Acros Supplier No. 295570010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-92-5 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYSA-N
Synonym camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
PubChem CID 6616
ChEBI CHEBI:3830
IUPAC Name 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
SMILES CC1(C2CCC(C2)C1=C)C

Specifications

Melting Point 42°C
Color White
Density 0.85
Boiling Point 159.0°C to 160.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Quantity 1 L
Beilstein 05, 156
Merck Index 15, 1732
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ether,cyclohexane,cyclohexene,,dioxane and chloroform,slightly soluble in alcohol
Viscosity 1.6 mPa.s (50°C)
Formula Weight 136.24
Percent Purity 75%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Camphene, 75%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Camphene, remainder mainly a-fenchene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.