missing translation for 'onlineSavingsMsg'
Learn More

Camphene, 75%, remainder mainly α-fenchene, Thermo Scientific Chemicals

Catalog Number 10351054
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
79-92-5
C10H16
136.24
MFCD00066603
CRPUJAZIXJMDBK-UHFFFAOYSA-N
camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
6616
CHEBI:3830
3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
CC1(C2CCC(C2)C1=C)C
This item is not returnable. View return policy
79-92-5
C10H16
136.24
MFCD00066603
CRPUJAZIXJMDBK-UHFFFAOYSA-N
camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
6616
CHEBI:3830
3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
CC1(C2CCC(C2)C1=C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 42°C
Color White
Density 0.85
Boiling Point 159.0°C to 160.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Quantity 1 L
Beilstein 05, 156
Merck Index 15, 1732
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ether,cyclohexane,cyclohexene,,dioxane and chloroform,slightly soluble in alcohol
Viscosity 1.6 mPa.s (50°C)
Formula Weight 136.24
Percent Purity 75%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Camphene, 75%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Camphene, remainder mainly a-fenchene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'