missing translation for 'onlineSavingsMsg'
Learn More

Campesterol, MedChemExpress

Product Code. 16470386
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16470386

Brand: Medchem Express HYN1459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Campesterol is a plant sterol with cholesterol lowering and anticarcinogenic effects.

Campesterol is a plant sterol with cholesterol lowering and anticarcinogenic effects.

Specifications

Chemical Name or Material Campesterol
CAS 474-62-4
Color White
Purity Grade Notes Research
Molecular Formula C28H48O
Quantity 2 mg
Synonym (24R)-5-Ergosten-3β-ol
Solubility Information Ethanol : 9.09 mg/mL (22.69 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.50 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 0.67 mg/mL (1.67 mM; Need ultrasonic)
SMILES C[C@@H](C(C)C)CC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molecular Weight (g/mol) 400.68
Formula Weight 400.68
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.