missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ cAMP-Screen™ Wash Buffer
Click to view available options
Quantity:
1 L
Unit Size:
Each
Description
- Quantity: 1L
Specifications
Specifications
| Product Type | Wash Buffer |
| Quantity | 1 L |
| Form | Liquid |
| For Use With (Equipment) | cAmp-Screen Direct™ Immunoassay System., cAMP-Screen™ Immunoassay System |
| Product Line | cAMP-Screen™ |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction