missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ cAMP-Screen Direct™ Cyclic AMP Immunoassay System

Product Code. 13305374
Click to view available options
Quantity:
192 assays (2 x 96)
960 assays
Unit Size:
Pack of 192
Pack of 960
This item is not returnable. View return policy

Product Code. 13305374

Brand: Applied Biosystems™ 4412187

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultrasensitive determination of cyclic AMP (cAMP) levels in cell lysates with a simplified protocol. Thermo Scientific cAMP-Screen Direct 96-well Cyclic AMP Immunoassay System is a competitive chemiluminescent immunoassay formatted with maximum flexibilit

  • Utilizes highly sensitive chemiluminescent CSPD™ Substrate with Sapphire-II™ Enhancer that is triggered by an enzyme conjugate composed of cAMP bound to alkaline phosphatase (cAMP-AP)
  • Ready-to-use chemiluminescent substrate/enhancer formulation generates sustained-glow light emission from 30 minutes after addition
  • Mainly used in secondary screening and pre-clinical research, where sensitivity and no false positives are essential
  • Highest sensitivity of any commercially available cAMP assay
  • Hours of read time with little or no degradation of signal
  • Simple protocol with cells grown directly on assay plate
  • Useful for wide range of receptor activation studies

High Sensitivity and Hours of Steady Glow Time
This chemiluminescent assay is designed to provide the highest sensitivity of any commercially available cAMP assay. As few as 60 femtomoles of cAMP can be detected. The assay has a wide dynamic range, detecting from 0.06 to 6,000 picomoles of cAMP without the need for sample dilution or manipulations such as acetylation. This is especially helpful in cell-based assays when measuring Gs- or Gi-coupled agonist or antagonist stimulation and/or inhibition. Intra-assay precision for duplicate samples is typically 5% or less. Once the substrate/enhancer formulation reaches glow signal, the plate can be read for hours with little or no degradation of the signal. This is useful in screens where several plates are compared with each other. In addition, the assay exhibits exceptionally low cross-reactivity with other adenosine-containing or cyclic nucleotides.

For Receptor Activation Studies
The cAMP-Screen system is designed for quantitation of cellular cAMP for functional assays of receptor activation. The assay has been used with established cell lines for functional measurements with endogenous receptors, cell lines with exogenously expressed ligand receptors on the cell surface, primary cell cultures, and tissues in response to treatment with the appropriate ligands. In addition it has been used for receptor characterization, orphan receptor ligand identification, and the characterization of novel chimeric receptors. The assay can be used for high-throughput screens for compounds that stimulate or interfere with these signal transduction pathways.

Specifications

  • Target: Cyclic AMP
  • Assay Incubation Time: 1 hr.
  • Assay Sensitivity: 60 femtomoles cAMP
  • Detection Method: Chemiluminescent
  • For use with Equipment: Microplate Reader
  • Label/Dye: Alkaline Phosphatase
  • Sample Type: Cell Lysates
  • Sample Volume: 60μL
  • Storage: Store at 2° to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 60 femtomoles cAMP
Content And Storage 1 kit, store at 2°C to 8 °C
Detection Method Chemiluminescence
Product Line cAMP-Screen Direct
Protein Family GPCR (G-Protein Coupled Receptors)
Sample Type Cell Culture
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Label or Dye AP
Quantity 960 assays
Sample Volume 60 μL
Target Kit Named Cyclic AMP
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.