missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ cAMP-Screen™ Assay/Lysis Buffer
Description
This Assay/Lysis Buffer is suitable for use with the cAMP-Screen™ or cAmp-Screen Direct™ Immunoassay System.
For Research Use Only. Not for use in diagnostics procedures.
- Quantity: 65 mL
Specifications
Specifications
| Product Type | ELISA Assay/Lysis Buffer |
| Quantity | 65 mL |
| Form | Liquid |
| For Use With (Equipment) | cAmp-Screen Direct™ Immunoassay System., cAMP-Screen™ Immunoassay System |
| Product Line | cAMP-Screen |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?