missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ cAMP Parameter Assay Kit

Product Code. p-4753494
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18752324

Brand: R&D Systems™ KGE002B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cAMP Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The cAMP Parameter Assay Kit quantifies multi-species cAMP.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.75 to 240 pmol/mL
Assay Sensitivity 8.57 pmol/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Specificity Cyclic AMP Levels in Various Samples
Sufficient For 1 x 96-well plate
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.