Learn More
CAMP/LL37/FALL39/Cathelicidin Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76864PCP
Product Code. 18503253
  • £452.00 / 0.10mL
Description
CAMP/LL37/FALL39/Cathelicidin Polyclonal antibody specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)