missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 RXFP3 (SALPR) Gi Cell Line

Product Code. 17334589
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17334589 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17334589 Supplier DiscoverX Supplier No. 950102C2
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The cAMP Hunter™ CHO-K1 RXFP3 (SALPR) Gi Cell Line measures RXFP3 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter RXFP3 (SALPR) Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. relaxin-3) based activation of RXFP3 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype RXFP3 , a naturally Gi/Go coupled GPCR. Upon activation of RXFP3 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Intracellular cAMP level
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist Recombinant Human Relaxin-3
Quantity 2 Vials
Target Entry RXFP3 (SALPR)
Accession Number NM_016568.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.