missing translation for 'onlineSavingsMsg'
Learn More

Camostat methanesulfate, 98%

Product Code. 15444759
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444759 500 mg 500mg
15434759 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444759 Supplier Thermo Scientific Alfa Aesar Supplier No. J62362.ME
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An orally active synthetic serine protease inhibitor | CAS: 59721-29-8 | C21H26N4O8S | 494.52 g/mol

It is used as a protease and Trypsin inhibitor. Furthermore it was found that this water induction and bicarbonate secretion increase was achieved via direct regulation of ductular cells of the dog pancreas. Other studies have shown Camostat mesylate to suppress spinal Fos expression in the rat, which is a marker for neuronal activation. In addition it was shown that this compound may prevent or depress pancreatitis-induced referred allodynia/hyperalgesia in rat models. Camostat mesylate has also demonstrated the ability to prevent Trypsin attack of microspheres, which causes the degradation product N-benzoyl-dl-arginine. Camostat mesylate is an inhibitor of KLK, plasminogen, Prothrombin and Trypsin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a protease and Trypsin inhibitor. Furthermore it was found that this water induction and bicarbonate secretion increase was achieved via direct regulation of ductular cells of the dog pancreas. Other studies have shown Camostat mesylate to suppress spinal Fos expression in the rat, which is a marker for neuronal activation. In addition it was shown that this compound may prevent or depress pancreatitis-induced referred allodynia/hyperalgesia in rat models. Camostat mesylate has also demonstrated the ability to prevent Trypsin attack of microspheres, which causes the degradation product N-benzoyl-dl-arginine. Camostat mesylate is an inhibitor of KLK, plasminogen, Prothrombin and Trypsin.

Solubility
Soluble in water (100 mM), DMSO (100 mM), and ethanol (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C21H26N4O8S
Molecular Weight (g/mol) 494.52
MDL Number MFCD00941410
InChI Key FSEKIHNIDBATFG-UHFFFAOYSA-N
Synonym camostat mesylate, camostat mesilate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
PubChem CID 5284360
IUPAC Name 4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid
SMILES CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1

Specifications

Melting Point 164°C to 168°C
Color White
Quantity 500 mg
Merck Index 14,1728
Solubility Information Soluble in water (100 mM),DMSO (100 mM),and ethanol (<1mg/ml).
Formula Weight 494.52
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Camostat methanesulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.