missing translation for 'onlineSavingsMsg'
Learn More

Camostat mesylate, MedChemExpress

Product Code. 16496206
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496206 100 mg 100mg
16475718 10 mg 10mg
16401887 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496206 Supplier Medchem Express Supplier No. HY13512
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.

Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.

Specifications

Chemical Name or Material Camostat mesylate
Physical Form Solid
CAS 59721-29-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H26N4O8S
For Use With (Application) Cancer-Kinase/protease
Synonym Camostat mesilate FOY305 FOY-S980
Solubility Information DMSO : 125 mg/mL (252.77 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.11 mM)
SMILES O=C(OCC(N(C)C)=O)CC1=CC=C(OC(C2=CC=C(NC(N)=N)C=C2)=O)C=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 494.52
Formula Weight 494.52
Percent Purity 98.45%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.