missing translation for 'onlineSavingsMsg'
Learn More

Camobucol, MedChemExpress

Product Code. 16490556
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490556 20 mg 20mg
16438847 5 mg 5mg
16436258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16490556 Supplier Medchem Express Supplier No. HY14916

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Camobucol (AGIX 4207) is an orally active, phenolic antioxidant and anti-inflammatory compound with antirheumatic properties.

Camobucol (AGIX 4207) is an orally active, phenolic antioxidant and anti-inflammatory compound with antirheumatic properties.

Specifications

Chemical Name or Material Camobucol
CAS 216167-92-9
Color White
Purity Grade Notes Research
Molecular Formula C33H50O4S2
Quantity 20 mg
Synonym AGIX 4207
SMILES CC(C)(C)C1=CC(SC(C)(C)SC2=CC(C(C)(C)C)=C(OCC(O)=O)C(C(C)(C)C)=C2)=CC(C(C)(C)C)=C1O
Molecular Weight (g/mol) 574.88
Formula Weight 574.88
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.