Learn More
Invitrogen™ Human CAMKK1 (CAMKKA), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q8N5S9 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 84254 |
| Molecular Weight (g/mol) | 59.8 kDa |
| Name | Human CAMKK1 (CAMKKA), His Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | AI846603; alpha CaMKK; calcium calmodulin dependent protein kinase kinase alpha; calcium/calmodulin dependent protein kinase kinase 1; calcium/calmodulin-dependent protein kinase kinase 1; calcium/calmodulin-dependent protein kinase kinase 1, alpha; calcium/calmodulin-dependent protein kinase kinase alpha; CaM-kinase IV kinase; caM-kinase kinase 1; caM-kinase kinase alpha; Camkk; caMKK 1; caM-KK 1; caMKK alpha; CaM-KK alpha; CAMKK alpha protein; CAMKK1; CAMKKA; CaMKKalpha |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.