missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKK alpha Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42682APC
This item is not returnable.
View return policy
Description
CaMKK alpha Polyclonal antibody specifically detects CaMKK alpha in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative)
Specifications
| CaMKK alpha | |
| Polyclonal | |
| Immunohistochemistry, Immunoprecipitation, Western Blot (Negative) | |
| calcium/calmodulin-dependent protein kinase kinase 1, calcium/calmodulin-dependent protein kinase kinase 1, alpha, Calcium/calmodulin-dependent protein kinase kinase alpha, CaM-kinase IV kinase, CaM-kinase kinase 1, CaM-kinase kinase alpha, CaM-KK 1, CaMKK alpha, CaM-KK alpha, CAMKK alpha protein, CAMKKACaMKK 1, DKFZp761M0423, EC 2.7.11, EC 2.7.11.17, MGC34095 | |
| The immunogen for this product maps to a region between residue 150 and 200 of human calcium/calmodulin-dependent protein kinase kinase 1, alpha using the numbering given in entry NP_757343.1 (GeneID 84254). | |
| 0.1 mL | |
| Protein Kinase | |
| 84254 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido