missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKIV Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13263-1-AP-150UL
This item is not returnable.
View return policy
Description
CaM kinase IV (CaMKIV), in a calcium and calmodulin dependent manner, is implicated in phosphorylating numerous transcription factors including the CCAAT enhancer binding protein (C/EBP), serum response factor (SRF), and cyclic AMP response element binding protein (CBP). The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.Specifications
| CaMKIV | |
| Polyclonal | |
| Unconjugated | |
| CAMK4 | |
| CaM kinase GR, CaMK GR, CAMK4, CaMKIV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12326, 25050, 814 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08414, P13234, Q16566 | |
| CAMK4 | |
| CaMKIV Fusion Protein Ag3922 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction