missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKIV Antibody (162), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90734B
This item is not returnable.
View return policy
Description
CaMKIV Monoclonal specifically detects CaMKIV in Mouse samples. It is validated for ELISA.
Specifications
| CaMKIV | |
| Monoclonal | |
| Biotin | |
| brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CaMKIV (P08414; Met1-Tyr469). | |
| 0.1 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase | |
| 814 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 162 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction