missing translation for 'onlineSavingsMsg'
Learn More

CaMKIV Antibody (162), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18395051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395051 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395051 Supplier Novus Biologicals Supplier No. NBP290734AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CaMKIV Monoclonal specifically detects CaMKIV in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKIV
Applications ELISA
Classification Monoclonal
Clone 162
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CaMKIV (P08414; Met1-Tyr469).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 814
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.