missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKII Pan Recombinant Rabbit Monoclonal Antibody (23GB5920), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | CaMKII Pan |
|---|---|
| Clone | 23GB5920 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089499
![]() |
Invitrogen™
MA552950 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087542
|
Invitrogen™
MA552951 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaMKII Pan Recombinant Monoclonal antibody specifically detects CaMKII Pan in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CaMKII Pan | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P11798, P28652, Q13554, Q13555, Q13557, Q6PHZ2, Q923T9, Q9UQM7 | |
| 108058, 12322, 12323, 12325, 815, 816, 817, 818 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5920 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; alpha CaM kinase II; alpha-CaMKII; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; calcium/calmodulin-dependent protein kinase type II subunit delta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit alpha; CaM kinase II subunit beta; caM kinase II subunit delta; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK1; Camk2; Camk2a; Camk2b; Camk2d; Camk2g; CAMKA; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; CaMKIINalpha; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; Ck2b; KIAA0968; Kiaa4163; mKIAA0968; PK2CDD; PKCCD; proline rich calmodulin-dependent protein kinase; R74975; RATCAMKI | |
| A synthesized peptide derived from human CaMKII (1-88AA). | |
| Primary | |
| -20°C | |
| Camk2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title