missing translation for 'onlineSavingsMsg'
Learn More

CaMKII-IN-1, MedChemExpress

Product Code. 16499967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499967 5 mg 5mg
16477488 10 mg 10mg
16417456 100 mg 100mg
16413517 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499967 Supplier Medchem Express Supplier No. HY18271
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CaMKII-IN-1 is a potent and highly selective CaMKII inhibitor with IC50 of 63 nM; significantly high selectivity against CaMKIV, MLCK, p38a, Akt1, and PKC.

CaMKII-IN-1 is a potent and highly selective CaMKII inhibitor with IC50 of 63 nM

Specifications

Chemical Name or Material CaMKII-IN-1
CAS 1208123-85-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H30ClN5O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 54 mg/mL (98.52 mM)
SMILES O=S(C1=CC=CC(Cl)=C1C)(NC2=C(CN(CC3=CC=CC=C3)CC4)C4=NC(NCCC5=CC=CC=C5)=N2)=O
Molecular Weight (g/mol) 548.1
Formula Weight 548.1
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.