missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKII delta Monoclonal Antibody (K52040_13E3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556282
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: LKKPDGVKES TESSNTTIED EDVKARKQEI IKVTEQLIEA INNGDFEAYT KICDPGLTAF EPEALGNLVE GMDFHRFYFE
CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns.
Specifications
| CaMKII delta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13557 | |
| Camk2d | |
| Recombinant human CAMK2D (immunogen range is 321-400). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K52040_13E3 | |
| Unconjugated | |
| Camk2d | |
| [d]-CaMKII; 2810011D23Rik; 8030469K03Rik; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II delta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; calcium/calmodulin dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase II, delta; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II subunit delta; calmodulin-dependent protein kinase II-delta dash; CaM kinase II delta subunit; caM kinase II subunit delta; CAM2; CaMK II; CAMK1; Camk2; CAMK2B; Camk2d; CAMK2G; CAMKB; CAMKD; CAMKG; Camki; CaMK-II delta subunit; CaMK-II subunit delta; CaM-kinase II delta chain; Kiaa4163; RATCAMKI | |
| Mouse | |
| Protein A | |
| RUO | |
| 817 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction