missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKII beta Antibody (S05-9G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19590-50ul
This item is not returnable.
View return policy
Description
CaMKII beta Monoclonal antibody specifically detects CaMKII beta in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CaMKII beta | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-9G5 | |
| Unconjugated | |
| calcium/calmodulin-dependent protein kinase (CaM kinase) II beta, calcium/calmodulin-dependent protein kinase II beta, calcium/calmodulin-dependent protein kinase type II beta chain, calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II beta subunit, CaM kinase II subunit beta, CAM2CAMK2, CAMKB, CaMK-II subunit beta, CaM-kinase II beta chain, EC 2.7.11, EC 2.7.11.17, MGC29528, proline rich calmodulin-dependent protein kinase | |
| A synthetic peptide of human CaMKII beta (Uniprot # Q13554) | |
| 50 μg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 816 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction