missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CaMKII beta Polyclonal antibody specifically detects CaMKII beta in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CaMKII beta |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Simple Western 1:5, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | calcium/calmodulin-dependent protein kinase (CaM kinase) II beta, calcium/calmodulin-dependent protein kinase II beta, calcium/calmodulin-dependent protein kinase type II beta chain, calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II beta subunit, CaM kinase II subunit beta, CAM2CAMK2, CAMKB, CaMK-II subunit beta, CaM-kinase II beta chain, EC 2.7.11, EC 2.7.11.17, MGC29528, proline rich calmodulin-dependent protein kinase |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: TRNFSVGRQTTAPATMSTAASGTTMGLVEQAKSLLNKKADGVKPQTNSTKNSAAATSPKGTLPPAALEPQTTVIH |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?