missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMKID Antibody (OTI2C6), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72074AF700
This item is not returnable.
View return policy
Description
CAMKID Monoclonal specifically detects CAMKID in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CAMKID | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CAMK1D | |
| Full length recombinant protein of human CAMKID (NP_065130) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2C6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| calcium/calmodulin-dependent protein kinase ID, calcium/calmodulin-dependent protein kinase type 1D, CaM kinase I delta, CaM kinase ID, CaM-K1, CaMKI delta, CaM-KI delta, CaMKID, CaMKI-like protein kinase, CKLiKEC 2.7.11.17, EC 2.7.11 | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase, Signal Transduction | |
| 57118.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction