Learn More
Invitrogen™ CaMKI gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA528580
Descripción
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Mouse brain, Rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Pig (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
Especificaciones
| CaMKI gamma | |
| Polyclonal | |
| Unconjugated | |
| Camk1g | |
| calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171358, 215303, 57172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.84 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q7TNJ7, Q91VB2, Q96NX5 | |
| Camk1g | |
| Recombinant protein encompassing a sequence within the center region of human CaMKI gamma. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.