missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaMKI gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113240
This item is not returnable.
View return policy
Description
CaMKI gamma Polyclonal Antibody for Western Blot, IHC (P), ELISA
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
Specifications
| CaMKI gamma | |
| Polyclonal | |
| Unconjugated | |
| Camk1g | |
| calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57172 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96NX5 | |
| Camk1g | |
| Recombinant Human Calcium/calmodulin-dependent protein kinase type 1G protein (217-476AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction