missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKI gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57828
This item is not returnable.
View return policy
Description
CaMKI gamma Polyclonal specifically detects CaMKI gamma in Human samples. It is validated for Western Blot.
Specifications
| CaMKI gamma | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| calcium/calmodulin-dependent protein kinase IG, calcium/calmodulin-dependent protein kinase type 1G, CaM kinase I gamma, CaM kinase IG, CaMKI gamma, CaM-KI gamma, caMKIG, CaMK-like CREB kinase III, CLICKIII, dJ272L16.1, EC 2.7.11, EC 2.7.11.17, VWS1CLICK III | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Equine: 100%; Mouse: 100%; Rabbit: 100%; Bovine: 92%; Chicken: 92%; Pig: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96NX5 | |
| CAMK1G | |
| Synthetic peptides corresponding to CAMK1G(calcium/calmodulin-dependent protein kinase IG) The peptide sequence was selected from the middle region of CAMK1G. Peptide sequence KDFICHLLEKDPNERYTCEKALSHPWIDGNTALHRDIYPSVSLQIQKNFA. | |
| 100 μL | |
| Protein Kinase | |
| 57172 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction