missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAMK1 (CaMK1), His Tag Recombinant Protein

Product Code. 11595016
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11595016

Brand: Invitrogen™ PV4391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14012
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 0.5mM EDTA, 50% glycerol, 0.02% Triton X-100, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 8536
Molecular Weight (g/mol) 45.4 kDa
Name Human CAMK1 (CaMK1), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19
Common Name CaMKI
Gene Symbol CAMK1
KinaseFamily CAMK1 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical ADP accumulation
Kinase Group Ser⁄Thr Kinases: CAMK Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Adapta™
Target Entry CAMK1
Target Gene calcium⁄calmodulin-dependent protein kinase I
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.