Learn More
Motic™ C-Mount Camera Adapter for SMZ168 Stereo Zoom Trinocular Microscope Chip Sensor
Product Code. p-8006207
Click to view available options
Magnification Power:
0.5X
Diameter (Metric):
12.7, 16.9 mm
Unit Size:
Each
Product Code. 11844313
Brand: Motic™ SW0113N6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mount a digital C-mount camera with 1/3 in., 1/2 in., or 2/3 in. on board chip on your SMZ168 Trinocular
  • For use with SMZ168 Stereo Zoom Trinocular Microscope:
    • SW0113N6: Mount a digital C-mount camera with 1/2 in. and 2/3 in. on board chip for 0.5X magnification power
    • SW0113N7: Mount a digital C-mount camera with 1/3 in. on board chip for 0.45X magnification power
Specifications
Type Camera Adapter
Diameter (English) 1/2, 2/3 in.
Diameter (Metric) 12.7, 16.9 mm
For Use With (Equipment) SMZ168 Stereo Zoom Microscope
Magnification Power 0.5X
Quantity 1 Ea.