missing translation for 'onlineSavingsMsg'
Learn More

CaM Kinase II Rabbit anti-Human, Mouse, Rat, Clone: 9P2E2, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £425.00

Specifications

Antigen CaM Kinase II
Clone 9P2E2
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18301392
View Documents
Novus Biologicals
NBP3-15256-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18384526
View Documents
Novus Biologicals
NBP3-15256-100UL
100 μg
£425.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaM Kinase II Monoclonal antibody specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

CaM Kinase II
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
816
IgG
Affinity purified
9P2E2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Neuroscience, Signal Transduction
PBS, 0.05% BSA, 50% glycerol, pH7.3
A synthetic peptide corresponding to a sequence within amino acids 550-650 of human CaM Kinase II (Q13554).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.