missing translation for 'onlineSavingsMsg'
Learn More

CaM Kinase II gamma Antibody (6A4A10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£187.00 - £435.00

Specifications

Antigen CaM Kinase II gamma
Clone 6A4A10
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications Western Blot, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697195
View Documents
Novus Biologicals
NBP2-37269-0.025ml
0.025 mL
£187.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18114278
View Documents
Novus Biologicals
NBP2-37269
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CaM Kinase II gamma Monoclonal specifically detects CaM Kinase II gamma in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CaM Kinase II gamma
1 mg/mL
Western Blot, ELISA
Unconjugated
RUO
Human
calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678
CAMK2G
IgG1
Affinity Purified
6A4A10
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Mouse
Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway
PBS with 0.05% Sodium Azide
818
Purified recombinant fragment of human CAMK2G (AA: 322-481) expressed in E. Coli.
Primary
62.6 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.