missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase II delta Rabbit anti-Human, Mouse, Rat, Clone: S03-8A9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14913-100UL
This item is not returnable.
View return policy
Description
CaM Kinase II delta Monoclonal antibody specifically detects CaM Kinase II delta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CaM Kinase II delta | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| S03-8A9 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II delta subunit, CaM kinase II subunit delta, CAMKD, CaMK-II delta subunit, CaMK-II subunit delta, CaM-kinase II delta chain, DKFZp686G23119, DKFZp686I2288, EC 2.7.11, EC 2.7.11.17, MGC44911 | |
| A synthetic peptide of human CaM Kinase II delta (Uniprot#: Q13557) | |
| 100 ul | |
| Protein Kinase, Wnt Signaling Pathway | |
| 817 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction