missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase II alpha, p Thr286 Antibody (SR1853), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CaM Kinase II alpha |
|---|---|
| Clone | SR1853 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621254
|
Novus Biologicals
NBP3-22608-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637604
|
Novus Biologicals
NBP3-22608-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaM Kinase II alpha Monoclonal antibody specifically detects CaM Kinase II alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
| CaM Kinase II alpha | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
| A synthesized peptide derived from human phospho-CaM Kinase II alpha [Thr286] (Uniprot #: Q9UQM7, P11798, P11275) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1853 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 815 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title