missing translation for 'onlineSavingsMsg'
Learn More

CaM Kinase I Antibody [DyLight 488], Novus Biologicals™

Product Code. 30732081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732081 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732081 Supplier Novus Biologicals Supplier No. AF7899K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CaM Kinase I Polyclonal antibody specifically detects CaM Kinase I in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaM Kinase I
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318
Host Species Sheep
Immunogen E. coli-derived recombinant human CaM Kinase I alpha , Ser176-Arg254 (Ala224Ser), Accession # Q14012
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Protein Kinase, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8536
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.