missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase I Antibody (010), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90023UV
This item is not returnable.
View return policy
Description
CaM Kinase I Monoclonal specifically detects CaM Kinase I in Human samples. It is validated for ELISA.
Specifications
| CaM Kinase I | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 10 | |
| ELISA | |
| calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CaM Kinase I (NP_003647.1; Leu2-Leu370). | |
| 0.1 mL | |
| Neuroscience, Protein Kinase, Sensory Systems, Vision | |
| 8536 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction