missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase I Antibody (010), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90023AF594
This item is not returnable.
View return policy
Description
CaM Kinase I Monoclonal specifically detects CaM Kinase I in Human samples. It is validated for ELISA.
Specifications
| CaM Kinase I | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 10 | |
| ELISA | |
| calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CaM Kinase I (NP_003647.1; Leu2-Leu370). | |
| 0.1 mL | |
| Neuroscience, Protein Kinase, Sensory Systems, Vision | |
| 8536 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido