missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calycosin-7-O-β-D-glucoside, MedChemExpress
Description
Calycosin-7-O-β-D-glucoside is an isoflavone isolated from Astragali Radix. Calycosin-7-O-β-D-glucoside has variety of biological activities, such as neuroprotective, cardioprotection, anti-inflammation, and antioxidative stress effects.
Specifications
Specifications
| Chemical Name or Material | Calycosin-7-O-β-D-glucoside |
| CAS | 20633-67-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H22O10 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (224.01 mM) |
| SMILES | O=C1C2=CC=C(O[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO)C=C2OC=C1C4=CC(O)=C(OC)C=C4 |
| Molecular Weight (g/mol) | 446.4 |
| Formula Weight | 446.4 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?