missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calumenin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100420
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Calumenin.
Caluminin is a 315 amino acid Ca2+-binding member of the CREC, EF-hand protein family. Calumenin is a secreted protein t hat contains six Ca2+-binding (EF-hand) motifs and is expressed in the lumen of the endoplasmic reticulum (ER) and Golgi apparatus. In the presence of Ca2+, Calumenin interacts with serum amyloid P component (SAP) and, together, they may play a role in the immunological defense system and participate in amyloidosis, the pathological formation of amyloid deposits in different types of tissues. Calumenin has an inhibitory effect on the vitamin K-dependent g-carboxylation system which converts vitamin K-dependent proteins to Gla-containing proteins. Calumenin may also be involved in the pathophysiology of thrombosis and/or wound healing by acting in an autocrine or paracrine manner.
Specifications
| Calumenin | |
| Polyclonal | |
| Unconjugated | |
| Calu | |
| 9530075H20Rik; CALU; Calumenin; Cbp50; CBP-50; Crocalbin; crocalbin-like protein; IEF SSP 9302; multiple EF-hand protein; Rcn; reticulocalbin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12321, 813 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35887, O43852 | |
| Calu | |
| A synthesized peptide derived from human CALU(Accession O43852), corresponding to amino acid residues L118-D168. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction