missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsyntenin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99468-50ul
This item is not returnable.
View return policy
Description
Calsyntenin-1 Polyclonal antibody specifically detects Calsyntenin-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Calsyntenin-1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:3000 | |
| alcadein alpha 1, Alcadein-alpha, ALC-ALPHA, alcalpha1, alcalpha2, Alzheimer-related cadherin-like protein, cadherin-related family member 12, calsyntenin 1, calsyntenin-1, CDHR12, CS1, CSTN1, FLJ32258, KIAA0911alcadein-alpha, Non-classical cadherin XB31alpha, non-classical cadherin XB31alpha1, PIK3CD, XB31alpha | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Calsyntenin-1. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 22883 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction