missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsyntenin-1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97117AF405
This item is not returnable.
View return policy
Description
Calsyntenin-1 Polyclonal specifically detects Calsyntenin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Calsyntenin-1 | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| alcadein alpha 1, Alcadein-alpha, ALC-ALPHA, alcalpha1, alcalpha2, Alzheimer-related cadherin-like protein, cadherin-related family member 12, calsyntenin 1, calsyntenin-1, CDHR12, CS1, CSTN1, FLJ32258, KIAA0911alcadein-alpha, Non-classical cadherin XB31alpha, non-classical cadherin XB31alpha1, PIK3CD, XB31alpha | |
| Produced in rabbits immunized with purified, recombinant Calsyntenin-1 (NP_001009566.1; Met1-Thr859). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 22883 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction