missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calsequestrin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555568
This item is not returnable.
View return policy
Description
Immunogen sequence: EEGLNFPTYD GKDRVVSLSE KNFKQVLKKY DLLCLYYHEP VSSDKVTQKQ FQLKEIVLEL VAQVL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.
Specifications
| Calsequestrin 2 | |
| Polyclonal | |
| Unconjugated | |
| Casq2 | |
| AA033488; AW146219; calsequestrin; calsequestrin 2; calsequestrin 2 (cardiac muscle); calsequestrin 2, fast-twitch, cardiac muscle; calsequestrin, cardiac muscle isoform; Calsequestrin-2; cardCSQ; cardiac calsequestrin; CASQ2; cCSQ; Csq2; ESTM52; LOW QUALITY PROTEIN: calsequestrin-2; PDIB2; SR calcium binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14958 | |
| Casq2 | |
| Recombinant protein corresponding to Human Calsequestrin 2. Recombinant protein control fragment (Product #RP-94140). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction