missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsequestrin 2 Antibody (4E10), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Calsequestrin 2 Monoclonal antibody specifically detects Calsequestrin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Calsequestrin 2 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E10 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | calsequestrin 2 (cardiac muscle), calsequestrin 2, fast-twitch, cardiac muscle, Calsequestrin, cardiac muscle isoform, calsequestrin-2, FLJ26321, FLJ93514, PDIB2 |
| Host Species | Mouse |
| Immunogen | Recombinant human Calsequestrin 2 (20-399aa) purified from E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?