missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsequestrin 2 Antibody (4E10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74043UV
This item is not returnable.
View return policy
Description
Calsequestrin 2 Monoclonal specifically detects Calsequestrin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Calsequestrin 2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CASQ2 | |
| Recombinant human CASQ2 (20-399aa) purified from E. coli | |
| 100 μL | |
| Cardiovascular Biology, Signal Transduction | |
| 845 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 40000000000 | |
| Western Blot, ELISA | |
| calsequestrin 2 (cardiac muscle), calsequestrin 2, fast-twitch, cardiac muscle, Calsequestrin, cardiac muscle isoform, calsequestrin-2, FLJ26321, FLJ93514, PDIB2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human CASQ2 mAb, clone 4E10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CASQ2 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering