missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsenilin Rabbit anti-Human, Clone: 11, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90302-100UL
This item is not returnable.
View return policy
Description
Calsenilin Monoclonal specifically detects Calsenilin in Human samples. It is validated for ELISA.
Specifications
| Calsenilin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| ELISA 1:5000-1:10000 | |
| calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3 | |
| Recombinant Human Calsenilin Protein | |
| 100 μg | |
| Vision | |
| 30818 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction