missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsenilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Calsenilin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Calsenilin Polyclonal specifically detects Calsenilin in Mouse samples. It is validated for Western Blot.Specifications
| Calsenilin | |
| Polyclonal | |
| Rabbit | |
| NP_001104801 | |
| 30818 | |
| Synthetic peptide towards CSEN. Peptide sequence PMSMEDSSDSELELSTVRHQPEGLDQLQAQTKFTKKELQSLYRGFKNECP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3 | |
| KCNIP3 | |
| IgG | |
| 26 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title