missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calsenilin Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00359G
This item is not returnable.
View return policy
Description
Calsenilin Polyclonal specifically detects Calsenilin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Calsenilin | |
| Polyclonal | |
| Western Blot, ELISA | |
| calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3 | |
| Produced in rabbits immunized with purified, recombinant Calsenilin (Q9Y2W7-1; Met1-Ile256). | |
| 0.1 mL | |
| Vision | |
| 30818 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction